3,5-Bis(trifluoromethyl)benzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15460-14
A15460-06
CAAAAA15460-14EA
445.56
CAD
CAAAAA15460-14
CAAAA15460-03
CAAAA15460-06
3,5-Bis(trifluoromethyl)benzonitrile 97%
3,5-Bis(trifluoromethyl)benzonitrile
Purity: 97%
CAS No.: 27126-93-8
Molecular Formula: C9H3F6N
Formula Weight: 239.12
Flash Point: 72°(161°F)
Refractive Index: 1.4175
Melting Point: 16-20°
Boiling Point: 154-156°
Beilstein Reference: 3552650
UN No.: 3276
MDL No.: MFCD00000379
Formula:
C₉H₃F₆N MW: 239.12 g/mol Boiling Pt: 155 Melting Pt: 16 Density: 1.421 Flash Pt: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000379 CAS Number: 27126-93-8 EINECS: 248-240-7 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...