m-Toluenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Toluenesulphonyl chloride, Toluene-3-sulphonyl chloride, Toluene-m-sulphonyl chloride, 3-Toluenesulfonyl chloride, Toluene-m-sulfonyl chloride, Toluene-3-sulfonyl chloride
A15458-06
A15458-14
A15458-22
CAAAA15458-06EA
106.49
CAD
CAAAA15458-06
CAAAA15458-14
CAAAA15458-22
m-Toluenesulfonyl chloride 98%
m-Toluenesulphonyl chloride
Purity: 98%
CAS No.: 1899-93-0
Molecular Formula: C7H7ClO2S
Formula Weight: 190.65
Flash Point: 107°(224°F)
Refractive Index: 1.5480
Odor: Acrid
Boiling Point: 252-253°
Beilstein Reference: 2087865
UN No.: 3265
MDL No.: MFCD00051747
Formula:
C₇H₇ClO₂S Boiling Pt: 252…253 °C Melting Pt: 11.7 °C Density: 1.314 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051747 CAS Number: 1899-93-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...