Diiodomethane 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger

A15457-14 A15457-22 A15457-36 A15457-0B A15457-18 A15457-30
CAAAAA15457-14EA 66.36 CAD
CAAAAA15457-14 CAAAAA15457-22 CAAAAA15457-36 CAAAA15457-0B CAAAA15457-18 CAAAA15457-30
Diiodomethane 99% stabilized
Diiodomethane

Purity: 99% CAS No.: 75-11-6 Molecular Formula: CH2I2 Formula Weight: 267.84 Flash Point: None Physical Form: Clear liquid Density: 3.322 Refractive Index: 1.7400 Sensitivity: Light Sensitive Appearance: Yellow Melting Point: 5-6° Boiling Point: 180-181° dec. Beilstein Reference: 1696892 Merck Reference: 14,6066 UN No.: 2810 MDL No.: MFCD00001079

Formula: CH₂I₂
Boiling Pt: 180…181 °C (dec.)
Melting Pt: 5…6 °C
Density: 3.322
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001079
CAS Number: 75-11-6
EINECS: 200-841-5
UN: 2810
ADR: 6.1,III
Merck Index: 14,6066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e37a85a8930c2cbeaf62626d427944a69722cc72&vid=73a1847aa5acf8fa3b0e777fe1bcae9e4f7bde3d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR