Bromocyclopentane 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15455-22 A15455-36
CAAAA15455-22EA 84.06 CAD
CAAAA15455-22 CAAAA15455-36
Bromocyclopentane 98+%
Bromocyclopentane

Purity: 98+% CAS No.: 137-43-9 Molecular Formula: C5H9Br Formula Weight: 149.04 Flash Point: 42°(107°F) Refractive Index: 1.4881 Odor: Odorless Boiling Point: 137-139° Beilstein Reference: 1209256 UN No.: 1993 MDL No.: MFCD00001359

Formula: C₅H₉Br
Boiling Pt: 137…139 °C
Density: 1.390
Flash Pt: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001359
CAS Number: 137-43-9
EINECS: 205-294-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR