2,5-Dichloropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15442-14
A15442-18
CAAAAA15442-14EA
173.8
CAD
CAAAAA15442-14
CAAAA15442-18
CAAAA15442-30
2,5-Dichloropyridine 98%
2,5-Dichloropyridine
Purity: 98%
CAS No.: 16110-09-1
Molecular Formula: C5H3Cl2N
Formula Weight: 147.99
Flash Point: 112°(233°F)
Odor: Unpleasant
Boiling Point: 190-191°
Beilstein Reference: 108886
UN No.: 2811
MDL No.: MFCD00006239
Formula:
C₅H₃Cl₂N Boiling Pt: 190…191 °C Melting Pt: 59…62 °C Flash Pt: 112 °C(233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006239 CAS Number: 16110-09-1 EINECS: 240-278-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...