7-Methoxycoumarin 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 7-Methoxy-2-benzopyrone, 7-Methoxy-2H-chromen-2-one

A15441-06 A15441-14 A15441-22
CAAAA15441-06EA 62.41 CAD
CAAAA15441-06 CAAAA15441-14 CAAAA15441-22
7-Methoxycoumarin 98+%
7-Methoxycoumarin

Purity: 98+% CAS No.: 531-59-9 Molecular Formula: C10H8O3 Formula Weight: 176.17 Melting Point: 117-120° Beilstein Reference: 141728

Formula: C₁₀H₈O₃
MW: 176.17 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 117…120 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006876
CAS Number: 531-59-9
EINECS: 208-513-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR