2-Cyclohexen-1-one 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15435-14
A15435-22
A15435-36
CAAAA15435-14EA
68.57
CAD
CAAAA15435-14
CAAAA15435-22
CAAAA15435-36
2-Cyclohexen-1-one 97%
2-Cyclohexen-1-one
Purity: 97%
CAS No.: 930-68-7
Molecular Formula: C6H8O
Formula Weight: 96.13
Flash Point: 56°(132°F)
Refractive Index: 1.4880
Melting Point: -53°
Boiling Point: 167-169°
Beilstein Reference: 1280477
UN No.: 2929
MDL No.: MFCD00001577
Formula:
C₆H₈O MW: 96.13 g/mol Boiling Pt: 167…169 °C Melting Pt: –53 °C Density: 0.993 Flash Pt: 56 °C (132 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001577 CAS Number: 930-68-7 EINECS: 213-223-5 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...