N-Acetyl-L-cysteine 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Acetylcysteine, N-Acetyl-3-mercaptoalanine, NAC, (R)-(+)-N-Acetylcysteine, (R)-2-Acetamido-3-mercaptopropanoic acid, Ac-Cys-OH, N-Acetyl-L-Cysteine, LNAC

A15409-14 A15409-22 A15409-36
CAAAA15409-14EA 109.23 CAD
CAAAA15409-14 CAAAA15409-22 CAAAA15409-36
N-Acetyl-L-cysteine 98+%
N-Acetyl-L-cysteine

A mucolytic agent for isolation of mycobacteria from sputum


Purity: 98+% CAS No.: 616-91-1 Molecular Formula: C5H9NO3S Formula Weight: 163.20 Melting Point: 108-111° Beilstein Reference: 1724426 Merck Reference: 14,88 Optical Rotation: +5.3° (c=2 in water) MDL No.: MFCD00004880


A mucolytic agent for isolation of mycobacteria from sputum

Formula: C₅H₉NO₃S
MW: 163.2 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 108…111 °C
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004880
CAS Number: 616-91-1
EINECS: 210-498-3
Merck Index: 14,88

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR