4-Heptylphenol 98+%

Supplier: Thermo Scientific Chemicals
Danger

A15407-03 A15407-06 A15407-14
CAAAA15407-03EA 72.68 CAD
CAAAA15407-03 CAAAA15407-06 CAAAA15407-14
4-Heptylphenol 98+%
4-Heptylphenol

Purity: 98+% CAS No.: 1987-50-4 Molecular Formula: C13H20O Formula Weight: 192.30 Refractive Index: 1.5070 Melting Point: 25-28° Boiling Point: 156°/9mm UN No.: 2430 MDL No.: MFCD00041751

Formula: C₁₃H₂₀O
Boiling Pt: 156 °C/9 mm
Melting Pt: 25…28 °C
Density: 0.939 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041751
CAS Number: 1987-50-4
EINECS: 217-862-0
UN: 2430
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR