4-Methylvaleric acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isocaproic acid, 4-Methylpentanoic acid
A15405-06
A15405-14
A15405-22
CAAAA15405-06EA
46.29
CAD
CAAAA15405-06
CAAAA15405-14
CAAAA15405-22
4-Methylvaleric acid 99%
4-Methylvaleric acid
Purity: 99%
CAS No.: 646-07-1
Molecular Formula: (CH3)2CH(CH2)2CO2H
Formula Weight: 116.16
Flash Point: 97°(206°F)
Refractive Index: 1.4146
Odor: Unpleasant
Melting Point: -35°
Boiling Point: 199-201°
Beilstein Reference: 1741912
UN No.: 2922
MDL No.: MFCD00002803
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 199…201 °C Melting Pt: –35 °C Density: 0.923 Flash Pt: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002803 CAS Number: 646-07-1 EINECS: 211-464-0 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...