1-Chloro-2-nitrobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Chloro-2-nitrobenzene, 2-Chloronitrobenzene
A15403-0B
A15403-0I
A15403-30
CAAAA15403-0BEA
117.08
CAD
CAAAA15403-0B
CAAAA15403-0I
CAAAA15403-30
1-Chloro-2-nitrobenzene 99%
1-Chloro-2-nitrobenzene
Purity: 99%
CAS No.: 88-73-3
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Flash Point: 126°(258°F)
Melting Point: 31-34°
Boiling Point: 245-246°
Beilstein Reference: 509273
Merck Reference: 14,2151
UN No.: 1578
MDL No.: MFCD00007061
Formula:
C₆H₄ClNO₂ Boiling Pt: 245…246 °C Melting Pt: 31…34 °C Density: 1.348 Flash Pt: 126 °C (258 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007061 CAS Number: 88-73-3 EINECS: 201-854-9 UN: 1578 ADR: 6.1,II Merck Index: 14,2151 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...