2,6-Dibromopyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15397-36
A15397-14
A15397-22
CAAAAA15397-36EA
1162.88
CAD
CAAAAA15397-36
CAAAA15397-14
CAAAA15397-22
2,6-Dibromopyridine 98%
2,6-Dibromopyridine
Purity: 98%
CAS No.: 626-05-1
Molecular Formula: C5H3Br2N
Formula Weight: 236.90
Flash Point: 213°(415°F)
Odor: Recognizable
Boiling Point: 255°
Beilstein Reference: 108922
UN No.: 2811
MDL No.: MFCD00006223
Formula:
C₅H₃Br₂N Boiling Pt: 255 °C Melting Pt: 117…120 °C Flash Pt: 213 °C (415 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006223 CAS Number: 626-05-1 EINECS: 210-926-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...