2-Chloropyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15394-09
A15394-14
A15394-22
CAAAAA15394-09EA
103.49
CAD
CAAAAA15394-09
CAAAA15394-14
CAAAA15394-22
2-Chloropyrimidine 98%
2-Chloropyrimidine
Purity: 98%
CAS No.: 1722-12-9
Molecular Formula: C4H3ClN2
Formula Weight: 114.54
Flash Point: 98°(208°F)
Melting Point: 64-67°
Boiling Point: 75-76°/10mm
Beilstein Reference: 107171
UN No.: 2811
MDL No.: MFCD00006060
Formula:
C₄H₃ClN₂ Boiling Pt: 75…76 °C/10 mm Melting Pt: 64…67 °C Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006060 CAS Number: 1722-12-9 EINECS: 217-020-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...