3,4-Dimethylbenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
A15388-06
A15388-14
CAAAA15388-06EA
139.67
CAD
CAAAA15388-06
CAAAA15388-14
3,4-Dimethylbenzaldehyde 97%
3,4-Dimethylbenzaldehyde
Purity: 97%
CAS No.: 5973-71-7
Molecular Formula: C9H10O
Formula Weight: 134.18
Refractive Index: 1.5510
Odor: Almond-like
Boiling Point: 226°
Beilstein Reference: 1634270
MDL No.: MFCD00016612
Formula:
C₉H₁₀O Boiling Pt: 226 °C Density: 1.017 Storage Temperature: Ambient |
MDL Number:
MFCD00016612 CAS Number: 5973-71-7 EINECS: 227-770-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...