4-Fluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15383-14
A15383-22
A15383-36
CAAAA15383-14EA
71.42
CAD
CAAAA15383-14
CAAAA15383-22
CAAAA15383-36
4-Fluorobenzaldehyde 98%
4-Fluorobenzaldehyde
Purity: 98%
CAS No.: 459-57-4
Molecular Formula: C7H5FO
Formula Weight: 124.11
Flash Point: 56°(133°F)
Refractive Index: 1.5210
Melting Point: -10°
Boiling Point: 180-181°
Beilstein Reference: 385857
UN No.: 1989
MDL No.: MFCD00003378
Formula:
C₇H₅FO Boiling Pt: 180…181 °C Melting Pt: –10 °C Density: 1.175 Flash Pt: 56 °C (133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003378 CAS Number: 459-57-4 EINECS: 207-293-6 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...