2-Chlorothanesulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2-Chloroethyl)sulphonyl chloride, (2-Chloroethyl)sulfonyl chloride

A15377-22 A15377-09 A15377-18 A15377-30
CAAAAA15377-22EA 609.88 CAD
CAAAAA15377-22 CAAAA15377-09 CAAAA15377-18 CAAAA15377-30
2-Chlorothanesulphonyl chloride 98%
2-Chlorothanesulphonyl chloride

Purity: 98% CAS No.: 1622-32-8 Molecular Formula: C2H4Cl2O2S Formula Weight: 163.02 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4915 Boiling Point: 203° Beilstein Reference: 1751202 UN No.: 3390 MDL No.: MFCD00007461

Formula: C₂H₄Cl₂O₂S
MW: 163.02 g/mol
Boiling Pt: 203 °C
Melting Pt: 3 °C
Density: 1.560
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007461
CAS Number: 1622-32-8
EINECS: 216-594-1
UN: 2927
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR