ω-Bromo-4-nitroacetophenone 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone

A15374-09 A15374-18
CAAAA15374-09EA 97.8 CAD
CAAAA15374-09 CAAAA15374-18
ω-Bromo-4-nitroacetophenone 95%
ω-Bromo-4-nitroacetophenone

Purity: 95% CAS No.: 99-81-0 Molecular Formula: C8H6BrNO3 Formula Weight: 244.05 Melting Point: 95-98° Beilstein Reference: 393567 UN No.: 1759 MDL No.: MFCD00007356

Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007356
CAS Number: 99-81-0
EINECS: 202-789-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR