Tris(3-methylphenyl)phosphine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(m-tolyl)phosphine, Tri-m-tolylphosphine

A15372-03 A15372-06 A15372-14
CAAAA15372-03EA 34.44 CAD
CAAAA15372-03 CAAAA15372-06 CAAAA15372-14
Tris(3-methylphenyl)phosphine 98+%
Tris(3-methylphenyl)phosphine

Purity: 98+% CAS No.: 6224-63-1 Molecular Formula: C21H21P Formula Weight: 304.37 Melting Point: 99-101° Beilstein Reference: 658863

Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00008532
CAS Number: 6224-63-1
EINECS: 228-312-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR