4-Fluoro-3-nitroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15371-36
A15371-14
A15371-22
CAAAAA15371-36EA
587.76
CAD
CAAAAA15371-36
CAAAA15371-14
CAAAA15371-22
4-Fluoro-3-nitroaniline 98%
4-Fluoro-3-nitroaniline
Purity: 98%
CAS No.: 364-76-1
Molecular Formula: C6H5FN2O2
Formula Weight: 156.12
Flash Point: 91°(195°F)
Melting Point: 94-98°
Beilstein Reference: 2210199
MDL No.: MFCD00007833
Formula:
C₆H₅FN₂O₂ Melting Pt: 94…98 °C Density: 0.43 g/cm³ (20 °C) Flash Pt: 91 °C(195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007833 CAS Number: 364-76-1 EINECS: 206-665-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...