p-Anisaldehyde 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Methoxybenzaldehyde
A15364-18
A15364-0B
A15364-30
CAAAAA15364-18EA
47.08
CAD
CAAAAA15364-18
CAAAA15364-0B
CAAAA15364-30
p-Anisaldehyde 98%
p-Anisaldehyde
Purity: 98%
CAS No.: 123-11-5
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 116°(241°F)
Physical Form: Liquid
Density: 1.121
Refractive Index: 1.5730
Sensitivity: Air and Light Sensitive
Appearance: Colorless to pale yellow
Odor: Odorless
Melting Point: 0-2°
Boiling Point: 247-249°
Beilstein Reference: 471382
Merck Reference: 14,663
MDL No.: MFCD00003385
Formula:
C₈H₈O₂ Boiling Pt: 247…249 °C Melting Pt: 0…2 °C Density: 1.121 Flash Pt: 116 °C(241 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003385 CAS Number: 123-11-5 EINECS: 204-602-6 Merck Index: 14,663 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...