2-Iodomesitylene 98%
Supplier: Thermo Scientific Chemicals
A15360-06
A15360-14
CAAAAA15360-22EA
1891.35
CAD
CAAAAA15360-22
CAAAA15360-06
CAAAA15360-14
2-Iodomesitylene 98%
2-Iodomesitylene
Purity: 98%
CAS No.: 4028-63-1
Molecular Formula: C9H11I
Formula Weight: 246.09
Melting Point: 28-30°
Beilstein Reference: 1860064
Formula:
C₉H₁₁I Melting Pt: 28…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013707 CAS Number: 4028-63-1 EINECS: 223-709-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...