Dimethyl oxalate 99%

Supplier: Thermo Scientific Chemicals
Warning

A15359-0E A15359-22 A15359-36
CAAAA15359-0EEA 892.7 CAD
CAAAA15359-0E CAAAA15359-22 CAAAA15359-36
Dimethyl oxalate 99%
Dimethyl oxalate

Purity: 99% CAS No.: 553-90-2 Molecular Formula: C4H6O4 Formula Weight: 118.09 Flash Point: 75°(167°F) Melting Point: 50-54° Boiling Point: 162-163° Beilstein Reference: 1071744 Merck Reference: 14,6106 MDL No.: MFCD00008442

Formula: C₄H₆O₄
Boiling Pt: 162…163 °C
Melting Pt: 50…54 °C
Density: 1.148
Flash Pt: 75 °C(167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008442
CAS Number: 553-90-2
EINECS: 209-053-6
UN: 3261
ADR: 8,III
Merck Index: 14,6106

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR