2-Acetyl-1-methyl-1H-pyrrole 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole
A15353-22
A15353-06
A15353-14
CAAAAA15353-22EA
617.78
CAD
CAAAAA15353-22
CAAAA15353-06
CAAAA15353-14
2-Acetyl-1-methyl-1H-pyrrole 98%
2-Acetyl-1-methyl-1H-pyrrole
Purity: 98%
CAS No.: 932-16-1
Molecular Formula: C7H9NO
Formula Weight: 123.16
Flash Point: 68°(154°F)
Refractive Index: 1.5420
Odor: Pleasant
Boiling Point: 200-202°
Beilstein Reference: 111887
MDL No.: MFCD00003089
Formula:
C₇H₉NO MW: 123.15 g/mol Boiling Pt: 200…202 °C Density: 1.04 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003089 CAS Number: 932-16-1 EINECS: 213-247-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...