N,N-Dimethylformamide dimethyl acetal 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1-Dimethoxytrimethylamine, Esterate M, N,N-Dimethylformamide Dimethyl Acetal, 1,1-Dimethoxy-N,N-dimethylmethylamine, DMF-DMA

A15350-14 A15350-22 A15350-36
CAAAA15350-14EA 76.42 CAD
CAAAA15350-14 CAAAA15350-22 CAAAA15350-36
N,N-Dimethylformamide dimethyl acetal 97%
N,N-Dimethylformamide dimethyl acetal

Purity: 97% CAS No.: 4637-24-5 Molecular Formula: (CH3)2NCH(OCH3)2 Formula Weight: 119.16 Flash Point: 7°(45°F) Refractive Index: 1.3972 Melting Point: -85° Boiling Point: 102-104° UN No.: 1993 MDL No.: MFCD00008482

Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
Boiling Pt: 102…104 °C
Melting Pt: –85 °C
Density: 0.897
Flash Pt: 7 °C (45 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008482
CAS Number: 4637-24-5
EINECS: 225-063-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR