Ethyl-3-hydroxybenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxybenzoic acid ethyl ester, Ethyl 3-hydroxybenzoate

A15343-36 A15343-14 A15343-22
CAAAAA15343-36EA 756.82 CAD
CAAAAA15343-36 CAAAA15343-14 CAAAA15343-22
Ethyl-3-hydroxybenzoate 99%
Ethyl 3-hydroxybenzoate

Purity: 99% CAS No.: 7781-98-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 72-76° Boiling Point: 187-188°/31mm MDL No.: MFCD00002296

Formula: C₉H₁₀O₃
Boiling Pt: 187…188 °C/31 mm
Melting Pt: 72…76 °C
Density: 1.068 g/cm³ (131 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002296
CAS Number: 7781-98-8
EINECS: 231-951-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR