3,5-Dihydroxybenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15338-14
A15338-03
A15338-06
CAAAAA15338-14EA
1047.54
CAD
CAAAAA15338-14
CAAAA15338-03
CAAAA15338-06
3,5-Dihydroxybenzaldehyde 98%
3,5-Dihydroxybenzaldehyde
Purity: 98%
CAS No.: 26153-38-8
Molecular Formula: C7H6O3
Formula Weight: 138.12
Physical Form: Powder
Sensitivity: Air Sensitive
Beilstein Reference: 1930147
MDL No.: MFCD00016611
Formula:
C₇H₆O₃ MW: 138.12 g/mol Melting Pt: 158…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016611 CAS Number: 26153-38-8 EINECS: 247-479-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...