Coumarin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone

A15336-0B A15336-0I A15336-30
CAAAA15336-0BEA 244.9 CAD
CAAAA15336-0B CAAAA15336-0I CAAAA15336-30
Coumarin 98%
Coumarin

Purity: 98% CAS No.: 91-64-5 Molecular Formula: C9H6O2 Formula Weight: 146.15 Flash Point: 162°(323°F) Odor: Pleasant Boiling Point: 297-299° Beilstein Reference: 383644 Merck Reference: 14,2562 UN No.: 2811 MDL No.: MFCD00006850

Formula: C₉H₆O₂
Boiling Pt: 297…299 °C
Melting Pt: 68…71 °C
Density: 0.935
Flash Pt: 162 °C (323 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006850
CAS Number: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,2562

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR