Indole-3-carbaldehyd 99%
Supplier: Thermo Scientific Chemicals
A15330-14
A15330-22
CAAAA15330-14EA
130.19
CAD
CAAAA15330-14
CAAAA15330-22
Indole-3-carbaldehyd 99%
Indole-3-carbaldehyd
Purity: 99%
CAS No.: 487-89-8
Molecular Formula: C9H7NO
Formula Weight: 145.16
Flash Point: 240°(464°F)
Melting Point: 195-198°
Beilstein Reference: 114117
MDL No.: MFCD00005622
Formula:
C₉H₇NO Melting Pt: 195…198 °C Density: 1,34 g/cm³ (20 °C) Flash Pt: 240 °C(464 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005622 CAS Number: 487-89-8 EINECS: 207-665-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...