1-Methylnaphthalene 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Methylnaphthalene

A15327-22 A15327-36
CAAAA15327-22EA 49.61 CAD
CAAAA15327-22 CAAAA15327-36
1-Methylnaphthalene 96%
1-Methylnaphthalene

Purity: 96% CAS No.: 90-12-0 Molecular Formula: C11H10 Formula Weight: 142.20 Flash Point: 82°(179°F) Refractive Index: 1.6150 Melting Point: -22° Boiling Point: 240-243° Beilstein Reference: 506793 UN No.: 3082 MDL No.: MFCD00004034

Formula: C₁₁H₁₀
MW: 142.2 g/mol
Boiling Pt: 240…243 °C
Melting Pt: –22 °C
Density: 1.024
Flash Pt: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004034
CAS Number: 90-12-0
EINECS: 201-966-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR