N,N,N-Trimethylanilinium tribromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide
A15326-0B
A15326-18
A15326-30
CAAAAA15326-0BEA
1474.14
CAD
CAAAAA15326-0B
CAAAA15326-18
CAAAA15326-30
N,N,N-Trimethylanilinium tribromide 97%
N,N,N-Trimethylanilinium tribromide
Purity: 97%
CAS No.: 4207-56-1
Molecular Formula: C6H5N(CH3)3Br.Br2
Formula Weight: 375.93
Odor: Irritating
Melting Point: 112-117°
Beilstein Reference: 3919459
UN No.: 3261
MDL No.: MFCD00011789
Formula:
C₉H₁₄Br₃N MW: 375.93 g/mol Melting Pt: 112…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011789 CAS Number: 4207-56-1 EINECS: 224-127-8 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...