(4-Bromophenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid

A15325-14 A15325-22
CAAAA15325-06EA 57.67 CAD
CAAAA15325-06 CAAAA15325-14 CAAAA15325-22
(4-Bromophenyl)acetic acid 99%
(4-Bromophenyl)acetic acid

Purity: 99% CAS No.: 1878-68-8 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 115-119° Beilstein Reference: 972146

Formula: (C₆H₄Br)CH₂COOH
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00004342
CAS Number: 1878-68-8
EINECS: 217-523-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR