ω-Bromo-4-fluoroacetophenone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone

A15319-03 A15319-06 A15319-14
CAAAAA15319-03EA 47.56 CAD
CAAAAA15319-03 CAAAA15319-06 CAAAA15319-14 CAAAA15319-22
ω-Bromo-4-fluoroacetophenone 97%
ω-Bromo-4-fluoroacetophenone

Purity: 97% CAS No.: 403-29-2 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 150°/12mm Beilstein Reference: 637863 MDL No.: MFCD00040830

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 150 °C/12 mm
Melting Pt: 45…49 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00040830
CAS Number: 403-29-2
EINECS: 206-955-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR