3-Tolualdehyde 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with hydroquinone (HQ) 0.1%

A15313-06 A15313-14 A15313-22
CAAAA15313-06EA 52.93 CAD
CAAAA15313-06 CAAAA15313-14 CAAAA15313-22
3-Tolualdehyde 97% stabilized
3-Tolualdehyde

Purity: 97% CAS No.: 620-23-5 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 78°(172°F) Refractive Index: 1.5410 Odor: Almond-like Boiling Point: 198-200° Beilstein Reference: 741964 MDL No.: MFCD00003374

Formula: C₈H₈O
Boiling Pt: 198…200 °C
Density: 1.020
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003374
CAS Number: 620-23-5
EINECS: 210-632-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR