4-Hydroxychalcone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Hydroxyphenyl)-1-phenylprop-2-en-1-one, 4-Hydroxybenzylidenacetophenone

A15307-06 A15307-14
CAAAA15307-06EA 88.01 CAD
CAAAA15307-06 CAAAA15307-14 CAAAA15307-22
4-Hydroxychalcone 97%
4-Hydroxychalcone

Purity: 97% CAS No.: 20426-12-4 Molecular Formula: C15H12O2 Formula Weight: 224.26 Melting Point: 183-185° Beilstein Reference: 2049155

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00016488
CAS Number: 20426-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR