1-(4-Iodophenyl)pyrrole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15305-03
A15305-06
CAAAAA15305-14EA
1380.05
CAD
CAAAAA15305-14
CAAAA15305-03
CAAAA15305-06
1-(4-Iodophenyl)pyrrole 97%
1-(4-Iodophenyl)pyrrole
Purity: 97%
CAS No.: 92636-36-7
Molecular Formula: C10H8IN
Formula Weight: 269.09
Melting Point: 128-133°
Beilstein Reference: 7915717
Formula:
C₁₀H₈IN Melting Pt: 128…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052399 CAS Number: 92636-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...