4-Chlorocinnamic acid predominantly trans 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15304-14
A15304-22
CAAAA15304-14EA
105.39
CAD
CAAAA15304-14
CAAAA15304-22
CAAAA15304-36
4-Chlorocinnamic acid predominantly trans 99%
4-Chlorocinnamic acid
Purity: 99%
CAS No.: 1615-02-7
Molecular Formula: C9H7ClO2
Formula Weight: 182.61
Melting Point: 248-253°
Beilstein Reference: 1365308
Formula:
C₉H₇ClO₂ Boiling Pt: 311 °C (1013 hPa) Melting Pt: 248…253 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004396 CAS Number: 1615-02-7 EINECS: 216-564-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...