2-Fluorocinnamic acid predominantly trans 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15303-06 A15303-14
CAAAA15303-06EA 83.11 CAD
CAAAA15303-06 CAAAA15303-14
2-Fluorocinnamic acid predominantly trans 98+%
2-Fluorocinnamic acid

Purity: 98+% CAS No.: 451-69-4 Molecular Formula: C9H7FO2 Formula Weight: 166.15 Odor: Odorless Melting Point: 178-182° Beilstein Reference: 1863963 MDL No.: MFCD00004370

Formula: C₉H₇FO₂
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00004370
CAS Number: 451-69-4
EINECS: 207-195-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR