2-Bromonaphthalene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15300-06
A15300-14
A15300-22
CAAAA15300-06EA
70.78
CAD
CAAAA15300-06
CAAAA15300-14
CAAAA15300-22
2-Bromonaphthalene 98+%
2-Bromonaphthalene
Purity: 98+%
CAS No.: 580-13-2
Molecular Formula: C10H7Br
Formula Weight: 207.08
Flash Point: 112°(233°F)
Odor: Odorless
Boiling Point: 281-282°
Beilstein Reference: 1858110
Merck Reference: 14,1426
MDL No.: MFCD00004051
Formula:
C₁₀H₇Br Boiling Pt: 281…282 °C Melting Pt: 53…57 °C Density: 1.605 Flash Pt: 112 °C(233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004051 CAS Number: 580-13-2 EINECS: 209-452-5 Merck Index: 14,1426 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...