3-Pentanone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethyl ketone

A15297-AE A15297-0F A15297-AP
CAAAAA15297-AEEA 72.52 CAD
CAAAAA15297-AE CAAAA15297-0F CAAAA15297-AP
3-Pentanone 99%
3-Pentanone

Purity: 99% CAS No.: 96-22-0 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 7°(45°F) Refractive Index: 1.3920 Odor: Acetone-like Melting Point: -39° Boiling Point: 102-103° Beilstein Reference: 635749 Merck Reference: 14,3121 UN No.: 1156 MDL No.: MFCD00009320

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 102…103 °C
Melting Pt: –39 °C
Density: 0.812
Flash Pt: 7 °C (45 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009320
CAS Number: 96-22-0
EINECS: 202-490-3
UN: 1156
ADR: 3,II
Merck Index: 14,3121

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR