4'-Butylacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

A15296-06 A15296-14
CAAAA15296-06EA 95.75 CAD
CAAAA15296-06 CAAAA15296-14
4'-Butylacetophenone 97%
4'-Butylacetophenone

Purity: 97% CAS No.: 37920-25-5 Molecular Formula: C12H16O Formula Weight: 176.26 Flash Point: 110°(230°F) Boiling Point: 101-102°/1.5mm MDL No.: MFCD00017500

Formula: C₁₂H₁₆O
Boiling Pt: 101…102 °C/1.5 mm
Density: 0.957
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017500
CAS Number: 37920-25-5
EINECS: 253-715-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR