α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Di(trifluoromethyl)aniline, 3,5-Bis(trifluoromethyl)aniline

A15289-09 A15289-18
CAAAA15289-09EA 42.66 CAD
CAAAA15289-09 CAAAA15289-18
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine 98%
α,α,α,α',α',α'-Hexafluoro-3,5-xylidine

Purity: 98% CAS No.: 328-74-5 Molecular Formula: C8H5F6N Formula Weight: 229.13 Flash Point: 83°(181°F) Refractive Index: 1.4340 Odor: Recognizable Boiling Point: 75-78°/10mm Beilstein Reference: 654318 UN No.: 2810 MDL No.: MFCD00000394

Formula: C₈H₅F₆N
MW: 229.12 g/mol
Boiling Pt: 75…78 °C/10 mm
Melting Pt: 3 °C
Density: 1.473
Flash Pt: 83 °C(181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000394
CAS Number: 328-74-5
EINECS: 206-335-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR