Furfuryl acetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15285-14
A15285-22
CAAAA15285-14EA
48.51
CAD
CAAAA15285-14
CAAAA15285-22
CAAAA15285-36
Furfuryl acetate 97%
Furfuryl acetate
Purity: 97%
CAS No.: 623-17-6
Molecular Formula: C7H8O3
Formula Weight: 140.14
Flash Point: 65°(149°F)
Refractive Index: 1.4630
Boiling Point: 175-177°
Beilstein Reference: 116128
MDL No.: MFCD00003251
Formula:
C₇H₈O₃ Boiling Pt: 175…177 °C Density: 1.118 Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003251 CAS Number: 623-17-6 EINECS: 210-775-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...