L(+)-Potassium hydrogen tartrate 98+%
Supplier: Thermo Scientific Chemicals
A15281-0B
A15281-0I
A15281-30
CAAAA15281-0BEA
176.96
CAD
CAAAA15281-0B
CAAAA15281-0I
CAAAA15281-30
L(+)-Potassium hydrogen tartrate 98+%
L(+)-Potassium hydrogen tartrate
Purity: 98+%
CAS No.: 868-14-4
Molecular Formula: C4H5KO6
Formula Weight: 188.18
Melting Point: ca 267° dec.
Beilstein Reference: 6119985
Merck Reference: 14,7615
MDL No.: MFCD00065392
Formula:
KOOC(CHOH)₂COOH Melting Pt: ∼ 267 °C (dec.) Density: 1.954 Storage Temperature: Ambient |
MDL Number:
MFCD00065392 CAS Number: 868-14-4 EINECS: 212-769-1 Merck Index: 14,7615 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...