5-Methyl-1,3-cyclohexanedione 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methylcyclohexane-1,3-dione

A15280-03 A15280-06
CAAAA15280-03EA 53.56 CAD
CAAAA15280-03 CAAAA15280-06 CAAAA15280-14
5-Methyl-1,3-cyclohexanedione 98%
5-Methyl-1,3-cyclohexanedione

Purity: 98% CAS No.: 4341-24-6 Molecular Formula: C7H10O2 Formula Weight: 126.16 Melting Point: 126-130° Beilstein Reference: 1099692

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00010379
CAS Number: 4341-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR