4-Formylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Terephthalaldehydic acid, 4-Carboxybenzaldehyde
A15277-09
A15277-18
A15277-30
CAAAA15277-09EA
99.07
CAD
CAAAA15277-09
CAAAA15277-18
CAAAA15277-30
4-Formylbenzoic acid 98%
4-Formylbenzoic acid
Purity: 98%
CAS No.: 619-66-9
Molecular Formula: C8H6O3
Formula Weight: 150.13
Odor: Odorless
Melting Point: ca 245°
Beilstein Reference: 471734
MDL No.: MFCD00006951
Formula:
C₈H₆O₃ MW: 150.13 g/mol Boiling Pt: 306 °C (1013 hPa) Melting Pt: ∼ 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006951 CAS Number: 619-66-9 EINECS: 210-607-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...