α,α,α-Trifluoro-4-tolualdehyde 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(Trifluoromethyl)benzaldehyde, 1-Formyl-4-trifluoromethylbenzene, p-Trifluoromethylbenzaldehyde

A15276-06 A15276-14
CAAAA15276-06EA 86.27 CAD
CAAAA15276-06 CAAAA15276-14
α,α,α-Trifluoro-4-tolualdehyde 97%
α,α,α-Trifluoro-4-tolualdehyde

Purity: 97% CAS No.: 455-19-6 Molecular Formula: C8H5F3O Formula Weight: 174.12 Flash Point: 65°(149°F) Refractive Index: 1.4630 Melting Point: 1-2° Boiling Point: 80-81°/25mm Beilstein Reference: 1101680 MDL No.: MFCD00006952

Formula: C₈H₅F₃O
MDL Number: MFCD00006952
CAS Number: 455-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR