3-Bromo-p-anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-4-methoxybenzaldehyde

A15274-06 A15274-14
CAAAA15274-06EA 53.88 CAD
CAAAA15274-06 CAAAA15274-14 CAAAA15274-22
3-Bromo-p-anisaldehyde 98%
3-Bromo-p-anisaldehyde

Purity: 98% CAS No.: 34841-06-0 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 49-53° Boiling Point: 108-110°/1mm MDL No.: MFCD00016599

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 108…110 °C/1 mm
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00016599
CAS Number: 34841-06-0
EINECS: 252-241-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR