ADA 98+%
Supplier: Thermo Scientific Chemicals
A15267-14
A15267-22
A15267-36
CAAAA15267-14EA
58.3
CAD
CAAAA15267-14
CAAAA15267-22
CAAAA15267-36
ADA 98+%
ADA
Purity: 98+%
CAS No.: 26239-55-4
Molecular Formula: H2NCOCH2N(CH2CO2H)2
Formula Weight: 190.16
Melting Point: ca 218° dec.
Beilstein Reference: 1787181
Merck Reference: 14,147
MDL No.: MFCD00008031
Formula:
H₂NC(O)CH₂N(CH₂COOH)₂ Melting Pt: ∼ 218 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008031 CAS Number: 26239-55-4 EINECS: 247-530-0 Merck Index: 14,147 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...