Linoleic acid ≥75% in aqueous solution, Technical Grade
Supplier: Thermo Scientific Chemicals
Synonyms:
(9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
A15265-09
A15265-18
A15265-30
CAAAA15265-09EA
86.43
CAD
CAAAA15265-09
CAAAA15265-18
CAAAA15265-30
Linoleic acid ≥75% in aqueous solution, Technical Grade
Linoleic acid
Purity: 75%
CAS No.: 60-33-3
Molecular Formula: C18H32O2
Formula Weight: 280.45
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₈H₃₂O₂ MW: 280.45 g/mol Boiling Pt: 230 °C/16 mm Density: 0.902 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064241 CAS Number: 60-33-3 EINECS: 200-470-9 Merck Index: 14,5505 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...