3,4-Dimethoxybenzenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,4-Dimethoxybenzene-1-sulphonyl chloride, 3,4-Dimethoxybenzene-1-sulfonyl chloride
A15263-06
A15263-14
CAAAA15263-06EA
112.97
CAD
CAAAA15263-06
CAAAA15263-14
CAAAA15263-22
3,4-Dimethoxybenzenesulfonyl chloride 98%
3,4-Dimethoxybenzenesulphonyl chloride
Purity: 98%
CAS No.: 23095-31-0
Molecular Formula: C8H9ClO4S
Formula Weight: 236.68
Melting Point: 66-70°
Beilstein Reference: 2840170
UN No.: 3261
MDL No.: MFCD00051769
Formula:
C₈H₉ClO₄S MW: 236.68 Melting Pt: 76 Storage Temperature: Ambient |
MDL Number:
MFCD00051769 CAS Number: 23095-31-0 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...